Herbicide mix #3 (2 components), derivatised form
Lieferant: Restek
32059
REST32059EA 182.5 EUR
REST32059
Herbicide mix #3 (2 components), derivatised form
Standards Organische Standards
This mix contains two components.
Components are MCPA methyl ester, MCPP (mecoprop) methyl ester.
Contains: MCPA methyl ester (2436-73-9); MCPP (Mecoprop) methyl ester (2786-19-8)
Standards: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfies ISO requirements.