Herbicide mix #4 (8 components), derivatised form
Lieferant: Restek
This mix contains eight components.
Components are acifluorfen methyl ester, bentazon methyl ester, chloramben methyl ester, DCPA methyl ester (chlorthal-dimethyl), 3,5-dichlorobenzoic acid methyl ester, 4-nitroanisole, pentachloroanisole, picloram methyl ester.
Contains: Acifluorfen methyl ester (50594-67-7); Bentazon methyl ester (61592-45-8); Chloramben methyl ester (7286-84-2); DCPA methyl ester (Chlorthal-dimethyl) (1861-32-1); 3,5-Dichlorobenzoic acid methyl ester (2905-67-1); 4-Nitroanisole (100-17-4); Pentachloroanisole (1825-21-4); Picloram methyl ester (14143-55-6)
Standards: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfies ISO requirements.