Pesticide performance evaluation mix with surrogates (8 components)
Lieferant: Restek
32074
REST32074EA
50
EUR
REST32074
Pesticide performance evaluation mix with surrogates (8 components)
Standards
Organische Standards
This mix contains eight components.
Components are α-BHC (1 μg/ml), β-BHC (1 μg/ml), γ-BHC (1 μg/ml), 4,4'-DDT (10 μg/ml), decachlorobiphenyl (SS) (2 μg/ml), endrin (5 μg/ml), methoxychlor (25 μg/ml), 2,4,5,6-tetrachloro-m-xylene (2 μg/ml).
Standards: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfies ISO requirements.