508.1 calibration mix #1 (17 components)
Lieferant: Restek
32094
REST32094EA 126 EUR
REST32094
508.1 calibration mix #1 (17 components)
Standards Organische Standards
Consists of 17 components, those are aldrin, α-bhc, β-bhc, δ-bhc, γ-bhc , 4,4'-ddd, 4,4'-dde, 4,4'-ddt, dieldrin, endosulfan i, endosulfan ii, endosulfan sulfate, endrin, endrin aldehyde, heptachlor, heptachlor epoxide and methoxychlor.
17 components
Standards: Certified reference material (CRM) manufactured and QC-tested in ISO-accredited labs satisfies ISO requirements.