508.1 calibration mix #2 (11 components)
Lieferant: Restek
32095
REST32095EA
126
EUR
REST32095
508.1 calibration mix #2 (11 components)
Standards
Organische Standards
Consists of 11 components, those are chlorobenzilate, cis-chlordane, trans-chlordane, chloroneb, dcpa methyl ester , etridiazole, hexachlorobenzene, cis-permethrin*, trans-permethrin*, propachlor and trifluralin.
11 components, Contains 500 µg/ml total permethrin.
Standards: Certified reference material (CRM) manufactured and QC-tested in ISO-accredited labs satisfies ISO requirements.
Bestellinformation: Exact content of each isomer is listed on certificate of analysis.