Pesticide standard mix A (9 components)
Lieferant: Restek
32297
REST32297EA 53 EUR
REST32297
Pesticide standard mix A (9 components)
Standards Organische Standards
Consists of nine components, those are α-bhc, γ-bhc, 4,4'-ddd, 4,4'-ddt, dieldrin, endosulfan i, endrin, heptachlor and methoxychlor.
Standards: Certified reference material (CRM) manufactured and QC-tested in ISO-accredited labs satisfies ISO requirements.