Environmental standard, minnesota Ag list 1 pesticides mix B (6 components)
Lieferant: Restek
32407
REST32407EA 54 EUR
REST32407
Environmental standard, minnesota Ag list 1 pesticides mix B (6 components)
Standards Umweltanalyse-Standards
Contains: Chlorpyrifos, EPTC, fonofos, phorate, terbufos, triallate.
Standards: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfies ISO requirements.