Organophosphorus pesticide mix (16 components), european formulation
Lieferant: Restek
32418
REST32418EA 187.5 EUR
REST32418
Organophosphorus pesticide mix (16 components), european formulation
Standards Organische Standards
Contains 16 components, those are acephate, azinphos methyl, chlorpyrifos, demeton-s-methyl, dichlorvos, dimethoate, ethion, malathion, methamidophos, methidathion, omethoate, pirimiphos methyl, profenofos, prothiofos, pyrazophos and tolclofos-methyl.
Standards: Certified reference material (CRM) manufactured and QC-tested in ISO-accredited labs satisfies ISO requirements.