Chlorinated acid herbicide mix (2 components)
Lieferant: Restek
32429
REST32429EA 48 EUR
REST32429
Chlorinated acid herbicide mix (2 components)
Standards Organische Standards
Contains two components, those are 2,4-dichlorophenoxyacetic acid and 2,4,5-TP.
Standards: Certified reference material (CRM) manufactured and QC-tested in ISO-accredited labs satisfies ISO requirements.