Chlorinated acids by LC, mix B (8 components)
Lieferant: Restek
32430
REST32430EA
117
EUR
REST32430
Chlorinated acids by LC, mix B (8 components)
Standards
Organische Standards
Consists of eight components, those are 2,4-db, 3,5-dichlorobenzoic acid, dinoseb, MCPA, MCPP, 4-nitrophenol, pentachlorophenol and 2,4,5-t.
Standards: Certified reference material (CRM) manufactured and QC-tested in ISO-accredited labs satisfies ISO requirements.