531.2 carbamate pesticide calibration mixture (11 components)
Lieferant: Restek
32435
REST32435EA
100
EUR
REST32435
531.2 carbamate pesticide calibration mixture (11 components)
Standards
Organische Standards
This mix contains 11 components.
- Complete set of materials for N-methylcarbamoyloximes and N-methylcarbamates
- New mix satisfies latest update of EPA Method 531.2
- Formulated in acetonitrile for stability and convenience for LC analysis
Components are aldicarb, aldicarb sulphone, aldicarb sulphoxide, carbaryl, carbophuran, 3-hydroxycarbophuran, methiocarb, methomyl, 1-naphthol, oxamyl, propoxur (baygon).
Standards: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfies ISO requirements.