Method 551.1 pesticide/herbicide mix (16 components)
Lieferant: Restek
32438
REST32438EA
124
EUR
REST32438
Method 551.1 pesticide/herbicide mix (16 components)
Standards
Organische Standards
Contains 16 components, those are alachlor, atrazine, bromacil, cyanazine , endrin, endrin aldehyde, endrin ketone, γ-bhc, heptachlor, heptachlor epoxide, hexachlorobenzene, hexachlorocyclopentadiene, methoxychlor, metolachlor, simazine and trifluralin.
Standards: Certified reference material (CRM) manufactured and QC-tested in ISO-accredited labs satisfies ISO requirements.