515.4 calibration mix (16 components)
Lieferant: Restek
32443
REST32443EA 93.5 EUR
REST32443
515.4 calibration mix (16 components)
Standards Organische Standards
Contains 16 components, those are acifluorfen, bentazon, chloramben, 2,4-d, dalapon, 2,4-db, dcpa diacid, dicamba, 3,5-dichlorobenzoic acid, dichlorprop, dinoseb, pentachlorophenol, picloram, quinclorac, 2,4,5-t and 2,4,5-tp.
Standards: Certified reference material (CRM) manufactured and QC-tested in ISO-accredited labs satisfies ISO requirements.