Methylated 515.4 chlorinated acids mix (16 components)
Lieferant: Restek
32444
REST32444EA 178 EUR
REST32444
Methylated 515.4 chlorinated acids mix (16 components)
Standards Organische Standards
Mix contains Acifluorfen methyl ester, Bentazon methyl ester, Chloramben methyl ester, Dalapon methyl ester, 2,4-D methyl ester, 2,4-DB methyl ester, DCPA methyl ester (Chlorthal-dimethyl), Dicamba methyl ester, 3,5-Dichlorobenzoic acid methyl ester, Dichlorprop methyl ester, Dinoseb methyl ether, Pentachloroanisole, Picloram methyl ester, Quinclorac methyl ester, 2,4,5-T methyl ester, 2,4,5-TP.
Standards: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy your ISO requirements.