Pesticide surrogate mix (2 components)
Lieferant: Restek
32453
REST32453EA
40
EUR
REST32453
Pesticide surrogate mix (2 components)
Standards
Umweltanalyse-Standards
Contains two components, those are decachlorobiphenyl and 2,4,5,6-tetrachloro-m-xylene.
Standards: Certified reference material (CRM) manufactured and QC-tested in ISO-accredited labs satisfies ISO requirements.