Organochlorine pesticide resolution check mix (with surrogates) (22 components)
Lieferant: Restek
32454
REST32454EA
82.5
EUR
REST32454
Organochlorine pesticide resolution check mix (with surrogates) (22 components)
Standards
Umweltanalyse-Standards
Contains 22 components, those are aldrin, α-bhc, β-bhc, δ-bhc, γ-bhc, cis-chlordane, trans-chlordane, decachlorobiphenyl, dieldrin, 4,4'-ddd, 4,4'-dde, 4,4'-ddt, endosulfan i, endosulfan ii, endosulfan sulfate, endrin, endrin aldehyde, endrin ketone, heptachlor, heptachlor epoxide, methoxychlor and 2,4,5,6-tetrachloro-m-xylene.
Standards: Certified reference material (CRM) manufactured and QC-tested in ISO-accredited labs satisfies ISO requirements.