EFSA PAH4 Standard
Lieferant: Restek
32469
REST32469EA 80.5 EUR
REST32469
EFSA PAH4 Standard
Standards Organische Standards
  • European Food Safety Authority (EFSA) PAH4 compounds prepared in toluene solvent
  • High 1,000 µg/ml concentration—dilute as needed to economically prepare custom mixes
  • Low volatility—longer shelf life and more accurate results.
  • High miscibility—compatible with quechers solvents
  • Certified reference material (CRM) meets strict ISO quality requirements
  • Pair with Rxi-PAH GC column for easy separation of EFSA PAH4 compounds from common interferences in food and environmental samples: www.restek.com/rxi-pah
Four components