EFSA PAH4 Standard
Lieferant: Restek
32469
REST32469EA
80.5
EUR
REST32469
EFSA PAH4 Standard
Standards
Organische Standards
- European Food Safety Authority (EFSA) PAH4 compounds prepared in toluene solvent
- High 1,000 µg/ml concentration—dilute as needed to economically prepare custom mixes
- Low volatility—longer shelf life and more accurate results.
- High miscibility—compatible with quechers solvents
- Certified reference material (CRM) meets strict ISO quality requirements
- Pair with Rxi-PAH GC column for easy separation of EFSA PAH4 compounds from common interferences in food and environmental samples: www.restek.com/rxi-pah
Four components