GC multiresidue pesticide kit
Lieferant: Restek
32562
REST32562EA 2631 EUR
REST32562
GC multiresidue pesticide kit
Standards Lebensmittel-Standards
Accurately identify and quantify pesticide residues by GC-MS/MS in fruits, vegetables, botanicals, and herbals such as tea, ginseng, ginger, echinacea, and dietary supplements.
  • Formulated and grouped for maximum long-term stability
  • Quantitatively tested to confirm composition
  • Contains 1 ml each of these mixtures
GC multiresidue pesticide kit consist of GC multiresidue pesticide standard #1, GC multiresidue pesticide standard #2, GC multiresidue pesticide standard #3.
Standards: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfies ISO requirements. Comprehensive 203-compound kit covers food safety lists by the FDA, USDA.