GC multiresidue pesticide standard #1 (16 components)
Lieferant: Restek
32563
REST32563EA 239 EUR
REST32563
GC multiresidue pesticide standard #1 (16 components)
Standards Lebensmittel-Standards
Organophosphorus compounds that has accurately identify and quantify pesticide residues by GC-MS/MS in fruits, vegetables, botanicals, and herbals such as tea, ginseng, ginger, echinacea, and dietary supplements.
Contains: Azinphos ethyl, azinphos methyl, chlorpyrifos, chlorpyrifos methyl, diazinon, EPN, fenitrothion, isazophos, phosalone, phosmet, pirimiphos ethyl, pirimiphos methyl, pyraclofos, pyrazophos, pyridaphenthion, quinalphos.