GC multiresidue pesticide standard #4 (28 components)
Lieferant: Restek
32566
REST32566EA 388.5 EUR
REST32566
GC multiresidue pesticide standard #4 (28 components)
Standards Lebensmittel-Standards
Organonitrogen compounds that has accurately identify and quantify pesticide residues by GC-MS/MS in fruits, vegetables, botanicals, and herbals such as tea, ginseng, ginger, echinacea, and dietary supplements.
Contains: Acetochlor, alachlor, allidochlor, clomazone (command), cycloate, diallate(cis & trans), dimethachlor, diphenamid, fenpropathrin, fluquinconazole, flutolanil, linuron, metazachlor, methoxychlor, metolachlor, N-(2,4-dimethylphenyl)formamide, norflurazon, oxadiazon, pebulate, pretilachlor, prochloraz, propachlor, propanil, propisochlor, propyzamide, pyridaben, tebufenpyrad, triallate.
Standards: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfies ISO requirements.