GC multiresidue pesticide standard #5 (34 compounds)
Lieferant: Restek
32567
REST32567EA 454.5 EUR
REST32567
GC multiresidue pesticide standard #5 (34 compounds)
Standards Organische Standards
Accurately identify and quantify pesticide residues by GC-MS/MS in fruits, vegetables, botanicals, and herbals such as tea, ginseng, ginger, echinacea and dietary supplements.
  • Quantitatively tested to confirm composition, detailed support documentation provided
  • Organonitrogen compound
Formulated and grouped for maximum long-term stability* and well-balanced chromatographic performance.
Optimised multiresidue pesticide method is offered free of charge and downloadable XLS file includes conditions and transition tables.
Consists of 34 components, those are atrazine, bupirimate, captafol, captan, chlorfenapyr, cyprodinil, etofenprox, etridiazole, fenarimol, fipronil, fludioxonil, fluridone , flusilazole, flutriafol, folpet, hexazinone , iprodione, lenacil, mgk-264, myclobutanil, paclobutrazol, penconazole, procymidone, propargite, pyrimethanil, pyriproxyfen, tebuconazole, terbacil, terbuthylazine, triadimefon, triadimenol, tricyclazole, triflumizole and vinclozolin.
Standards: Certified reference material (CRM) manufactured and QC-tested in ISO-accredited labs satisfies ISO requirements.
Bestellinformation: The Comprehensive 203-compound kit also available (cat. # 32562) that covers food safety lists by the FDA, USDA, and other global governmental agencies; other ampoules also sold separately.
* NOTE: When combining many compounds with different chemical functionalities, mix stability can be an issue. In formulating these standards, we extensively studied the 203 compounds involved, and then grouped them into as few mixes as possible while still ensuring maximum long-term stability and reliability. For quantitative analysis, we recommend analysing each mix separately to ensure accurate results for every compound.