GC multi-residue pesticide standard #9 (8 components)
Lieferant: Restek
32571
REST32571EA 145.5 EUR
REST32571
GC multi-residue pesticide standard #9 (8 components)
Standards Organische Standards
The multiresidue accurately identify and quantify pesticide residues by gc-ms/ms in fruits, vegetables, botanicals, and herbals such as tea, ginseng, ginger, echinacea, and dietary supplements.
  • Quantitatively tested to confirm composition, detailed support documentation provided
  • Formulated and grouped for maximum long-term stability
Ampul 7 of 9 in the GC-MS/MS multiresidue pesticide standard kit.
The multiresidue pesticide standard consists of eight components disulphoton, fonofos, methyl parathion, mevinphos, parathion (ethyl parathion), phorate, piperonyl butoxide, triazophos.
Standards: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy ISO requirements.
Bestellinformation: NOTE: When combining a large number of compounds with different chemical functionalities, mix stability can be an issue. In formulating these standards, we extensively studied the 203 compounds involved, then grouped them into as few mixes as possible while still ensuring maximum long-term stability and reliability. For quantitative analysis, we recommend analyzing each mix separately to ensure accurate results for every compound.