Pesticides mix #2, method 527 (5 components)
Lieferant: Restek
33008
REST33008EA
107
EUR
REST33008
Pesticides mix #2, method 527 (5 components)
Standards
Organische Standards
The pesticides mix consists of five components chlorpyrifos, dimethoate, malathion, parathion and terbufos sulphone.
Standards: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy ISO requirements.