Oxy setup blend (29 components)
Lieferant: Restek
ASTM Method D6730-01 is specifically designed for the determination of the individual hydrocarbons present in spark ignition fuels, as well as fuel blends containing oxygenates such as methyl tert-butyl ether, ethyl tert-butyl ether, tert-butanol, ethanol, etc.
It consists of 29 components, they are benzene (1,00%), tert-butanol (0,50%), cyclohexane (28,9%), n-decane (1,00%), 2,3-dimethylbutane (0,50%), trans-1,2-dimethylcyclopentane (0,50%), 2,3-dimethylheptane, (0,20%), dodecane (0,25%), ethanol (8,00%), ethylbenzene (25,0%) ,3-ethylpentane (0,20%), n-heptane (2,00%), n-hexane (2,00%) ,2-methylbutene-2 (2,50%), methyl tert-butyl ether (10,0%), 1-methylcyclopentene (0,50%), 1-methyl-2-ethylbenzene (0,50%), 1-methylnaphthalene (0,25%), 5-methylnonane, (0,20%), naphthalene (0,50%), n-nonane (2,00%), n-octane (2,00%), n-pentane (2,00%), 1,2,3,5-tetramethylbenzene (0,25%), toluene (7,00%), tridecane (0,25%), 2,3,3-trimethylpentane (0,50%), undecane (0,50%), p-xylene (1,00%),