Sulphur simulated distillation standard (SSDS)
Lieferant: Restek
33049
REST33049EA
330
EUR
REST33049
Sulphur simulated distillation standard (SSDS)
Standards
Mineralölindustrie-Standards
The standard consists of ethanethiol, 1-propanethiol, 1-butanethiol, 1-pentanethiol, 1-hexanethiol, 1-heptanethiol, 3,5-dimethylbenzenethiol, 1-octanethiol, 1-nonanethiol, 1-decanethiol, 1-pentadecanethiolol, 1-hexadecanethiol and 1-octadecanethiol.