8270 Matrix Spike Mix (76 components)
Lieferant: Restek
31687 31687.15 33073
REST31687EA 279.5 EUR
REST31687 REST31687.15 REST33073
8270 Matrix Spike Mix (76 components)
Standards Organische Standards
The analysis of semivolatile organic pollutants in solid waste, soil, water, and air matrices using gas chromatography - mass spectrometry (GC - MS).
1,2-diphenylhydrazine decomposes to azobenzene (mix component) in the injector. N-nitrosodiphenylamine decomposes to diphenylamine (mix component) in the injector.
Standards: EPA standards.