CARB 1004 aldehyde/ketone-DNPH calibration standard (13 components)
Lieferant: Restek
33093
REST33093EA 114.5 EUR
REST33093
CARB 1004 aldehyde/ketone-DNPH calibration standard (13 components)
Standards Organische Standards
The CARB 1004 aldehyde/ketone-DNPH calibration standard consists of 13 components, they are acetaldehyde-2,4-DNPH, acetone-2,4-DNPH, acrolein-2,4-DNPH, benzaldehyde-2,4-DNPH, n-butyraldehyde-2,4-DNPH, crotonaldehyde-2,4-DNPH, formaldehyde-2,4-DNPH, hexaldehyde-2,4-DNPH, methacrolein-2,4-DNPH, methyl ethyl ketone-2,4-DNPH, propionaldehyde-2,4-DNPH, m-tolualdehyde-2,4-DNPH, valeraldehyde-2,4-DNPH.
Standards: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy ISO requirements.