SOM01.1 deuterated monitoring compound mix SIM compounds (2 components)
Lieferant: Restek
33913
REST33913EA 90 EUR
REST33913
SOM01.1 deuterated monitoring compound mix SIM compounds (2 components)
Standards Organische Standards
The deuterated monitoring compound mix consists of fluoranthene-d10 and 2-methylnaphthalene-d10.
Standards: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy ISO requirements.