Amine column test mix (GC) (8 components)
Lieferant: Restek
The amine column test mix consists of (C10) decane (0,60 mg/ml), (C12) dodecane (0.60 mg/ml, 1,2-butanediol (0.60 mg/ml), diethanolamine (DEA) (1.20 mg/ml), diethylenetriamine (1.20 mg/ml), 2,6-dimethylaniline (0.60 mg/ml), 2-nonanol (0.60 mg/ml), pyridine(0.60 mg/ml).
Standards: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy your ISO requirements.