LC reversed phase test mix #1 (4 components)
Lieferant: Restek
35005
REST35005EA
62
EUR
REST35005
LC reversed phase test mix #1 (4 components)
Standards
Organische Standards
Routine analysis using this mix can assist in determining the need to perform column and/or system maintenance.
The LC reversed phase test mix #1 consists of four components, they are benzene (3,00 mg/ml), biphenyl (0.06 mg/ml), naphthalene (0,50 mg/ml) and uracil (0,02 mg/ml).
Standards: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy ISO requirements.