FAME #3 mix
Lieferant: Restek
35012
REST35012EA 118 EUR
REST35012
FAME #3 mix
Standards Lebensmittel-Standards
This mixture can be used for quantification (AOCS method CE 1a–13) and approximate the compositions of oils.
  • Suitable for oils of short to mid-range chain lengths (C8 - C16)
This mix contains five components such as C8:00 methyl caprylate (20%), C10:00 methyl caprate (20%), C12:00 methyl laurate (20%), C14:00 methyl myristate (20%), C16:00 methyl palmitate (20%).