FAME #16 mix
Lieferant: Restek
35022
REST35022EA 118 EUR
REST35022
FAME #16 mix
Standards Lebensmittel-Standards
This mixture can be used for quantification (AOCS Method CE 1a–13) and approximate the compositions of the types of oils like corn, poppy seed, cotton seed, soybean, walnut, safflower, sunflower, rice, bran, and sesame oil.
This mix contains six components such as C16:00 methyl palmitate (6%), C18:00 methyl stearate (3%), C18:01 methyl oleate (35%), C18:02 methyl linoleate (50%), C18:03 methyl linolenate (3%), C20:00 methyl arachidate (3%).
% represents weight/weight% basis (minimum 50 mg/ampoule).