FAME #20 mix (8 components)
Lieferant: Restek
35026
REST35026EA
148
EUR
REST35026
FAME #20 mix (8 components)
Standards
Lebensmittel-Standards
This mixture can be used for quantification (AOCS Method CE 1a–13) and approximate the compositions of the types of oils like coconut, palm kernel, babassu, and ouri-curi oil.
This mix contains eight components such as C8:00 methyl caprylate (7%), C10:00 methyl caprate (5%), C12:00 methyl laurate (48%), C14:00 methyl myristate (15%), C16:00 methyl palmitate (7%), C18:00 methyl stearate (3%), C18:01 methyl oleate (12%), C18:02 methyl linoleate (3%).
% represents weight/weight% basis (minimum 50 mg/ampoule).