NPI-1000 column test mix (8 components)
Lieferant: Restek
35224
REST35224EA 67.5 EUR
REST35224
NPI-1000 column test mix (8 components)
Standards Gaschromatographie-Standards
The mix consists of (C9:0) methyl nonanoate, (C13) n-tridecane, (C15) n-pentadecane, acenaphthylene, 4-chlorophenol, 1,6-hexanediol, 4-propylaniline and 1-undecanol.
Standards: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy your ISO requirements.