Test mix for NPI-350 column (8 components)
Lieferant: Restek
This test mix for NPI-350 column contains: (C9:0 FAME) Methyl nonanoate (1731-84-6); (C13) n-Tridecane (629-50-5); (C15) n-Pentadecane (629-62-9); Acenaphthylene (208-96-8); 4-Chlorophenol (106-48-9); 1,6-Hexanediol (629-11-8); 4-Propylaniline (2696-84-6); 1-Undecanol (112-42-5).