Grob mix 280 (10 Components)
Lieferant: Restek
35227
REST35227EA
61
EUR
REST35227
Grob mix 280 (10 Components)
Standards
Organische Standards
The grob mix 280 consists of 10 components n-decane (280 μg/ml), methyl decanoate (420 μg/ml), n-undecane (280 μg/ml), methyl undecanoate (420 μg/ml), methyl dodecanoate (420 μg/ml), L-2,3-butanediol (530 μg/ml), 2,6-dimethylaniline (320 μg/ml), 2,6-dimethylphenol (320 μg/ml), nonanal (400 μg/ml) and 1-octanol (360 μg/ml).
Standards: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy your ISO requirements.