Phenols/anilines/pesticides test mix (9 components)
Lieferant: Restek
35245
REST35245EA 50 EUR
REST35245
Phenols/anilines/pesticides test mix (9 components)
Standards Organische Standards
The mix consists of nine components, they are 4-chloroaniline, 4,4'-DDT, 2,4-dinitrophenol, endrin, naphthalene, 3-nitroaniline, n-nitroso-di-n-propylamine, pentachlorophenol, phenanthrene.
Standards: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy ISO requirements.