Free fatty acids test standard (6 components)
Lieferant: Restek
35272
REST35272EA
64
EUR
REST35272
Free fatty acids test standard (6 components)
Standards
Organische Standards
The free fatty acids test standard consists of six components they are (C4:0) butyric acid, acetic acid, isobutyric acid, isovaleric acid, propionic acid and valeric acid.
Standards: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy ISO requirements.