Chlortrimethylsilan

Lieferant: Restek

Synonyme: TMCS, Trimethylsilylchlorid, Trimethylchlorsilan, TMSC

35611
REST35611EA 152 EUR
REST35611
Chlortrimethylsilan
Chlortrimethylsilan
Formel: C₃H₉ClSi
Molecular Weight: 108,64 g/mol
MDL: MFCD00000502
CAS-Nummer: 75-77-4
UN: 1298
ADR: 3,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR