N-Methylbistrifluoracetamid

Lieferant: Restek
Gefahr

Synonyme: 2,2,2-Trifluoro-N-methyl-N-(2,2,2-trifluoroacetyl)acetamide, MBTFA

35616
REST35616EA 0 EUR
REST35616
N-Methylbistrifluoracetamid
N-Methylbistrifluoracetamid

Acylation derivatization reagent.


  • Most commonly used for electron capture detection
  • React with alcohols, amines, and phenols
  • Frequently used for drugs of abuse confirmation


Acylation reagents offer the same advantage available from silylation reagents: creating less polar, more volatile derivatives. In comparison to silylating reagents, the acylating reagents can more readily target highly polar multifunctional compounds, such as carbohydrates and amino acids. In addition, acylating reagents offer the distinct advantage of introducing electron-capturing groups, thus enhancing detectability during analysis.

Formel: C₅H₃F₆NO₂
Molecular Weight: 223,07 g/mol
MDL: MFCD00000412
CAS-Nummer: 685-27-8
EINECS: 211-680-5
UN: 2920
ADR: 8.3,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR