European pharmacopoeia/ICH Class 2 mix B, revised (10 components)
Lieferant: Restek
36270
REST36270EA
76.5
EUR
REST36270
European pharmacopoeia/ICH Class 2 mix B, revised (10 components)
Standards
Organische Standards
The european pharmacopoeia/ICH Class 2 mix B, revised consists of 10 components, they are acetonitrile (410 μg/ml), chloroform (60 μg/ml), 1,2-dimethoxyethane (100 μg/ml), N,N-dimethylacetamide (1,090 μg/ml), 1,4-dioxane (380 μg/ml), 2-hexanone (50 μg/ml), methanol (3,000 μg/ml), nitromethane (50 μg/ml), pyridine (200 μg/ml) and tetralin (100 μg/ml).
Standards: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy ISO requirements.