Residual Solvents Class 2 - Mix B (8 components)
Lieferant: Restek
The solvents consists of (C6) n-hexane (290 μg/ml), chloroform (60 μg/ml), 1,2-dimethoxyethane (100 μg/ml), 2-hexanone (50 μg/ml), nitromethane (50 μg/ml), pyridine (200 μg/ml), tetralin (100 μg/ml), trichloroethene (80 μg/ml).
Standards: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy your ISO requirements.