HPLC columns, Viva C8 (USP L7)
Lieferant: Restek
9513525 951350210 951350212 9513511 9513512 9513512-700 9513513-700 9513515 9513515-700 951351E 9513521 9513522 9513522-700 9513523-700 9513525-700 951352E 9513531 9513532-700 9513533-700 9513535 9513535-700 951353E 9513551 9513552 9513552-700 9513553-700 9513555 9513555-700 951355E 9513561 9513562 9513562-700 9513563-700 9513565 9513565-700 951356E 9513571 9513572 9513572-700 9513573-700 9513575 9513575-700 9513532 951357E
REST9513525EA 1389 EUR
REST9513525 REST951350210 REST951350212 REST9513511 REST9513512 REST9513512-700 REST9513513-700 REST9513515 REST9513515-700 REST951351E REST9513521 REST9513522 REST9513522-700 REST9513523-700 REST9513525-700 REST951352E REST9513531 REST9513532-700 REST9513533-700 REST9513535 REST9513535-700 REST951353E REST9513551 REST9513552 REST9513552-700 REST9513553-700 REST9513555 REST9513555-700 REST951355E REST9513561 REST9513562 REST9513562-700 REST9513563-700 REST9513565 REST9513565-700 REST951356E REST9513571 REST9513572 REST9513572-700 REST9513573-700 REST9513575 REST9513575-700 REST9513532 REST951357E
HPLC columns, Viva C8 (USP L7)
Chromatographiesäulen
Viva C8 columns are conventional monomeric octylsilane columns offering a shorter alkyl chain to provide less hydrophobic retention and improved basic peak shape over a traditional C18 phase. These general-purpose C8 columns are suitable for a wide range of compounds from acidic through slightly basic.
  • Rugged, spherical particles with 300 Å pore size
  • High proportion of pore/surface area available to large molecules
  • Excellent for separating peptides or proteins