Restek's FluoroPhenyl phase offers chromatographers the ability to run in reversed-phase or HILIC mode for a variety of compounds.
- Capable of both reversed-phase and HILIC separations
- Ideal for increasing sensitivity and selectivity in LC-MS analyses
- Offers increased retention for charged bases
The culmination of 20+ years in LC, Force performance LC columns are long-lasting and reproducible, even under the stress of UHPLC.
Restek's FluoroPhenyl phase offers chromatographers the ability to run in reversed-phase or HILIC mode for a variety of compounds. Restek's FluoroPhenyl columns are also amenable to LC-MS because they are extremely reliable and efficient with acidic mobile phases.
Premium quality is backed by the strength of our 100% Pure Satisfaction guarantee. And perhaps most important, Force columns are fully scalable between 3 or 5 µm HPLC and 1,8 µm UHPLC, so you can easily transfer and optimize methods without extensive calculations.
Bestellinformation: Switch to a FluoroPhenyl LC column when limited retention and selectivity are observed on a C18 for basic compounds or when you need increased retention of hydrophilic compounds.