Vibratory sieve shaker, AS 450 control
Lieferant: Retsch GmbH
30.026.0001
RETS30.026.0001EA 9190 EUR
RETS30.026.0001
Vibratory sieve shaker, AS 450 control
Schüttler Siebmaschinen
The analytical sieve shaker AS 450 control is used in research and development, quality control of raw materials, interim and finished products as well as in production monitoring. The controllable electromagnetic drive offers an optimal adaption for every product. Sharp fractions are obtainable even after very short
sieving times. It is suitable for dry and wet sieving.
  • Easy operation, ergonomic design and maintenance-free
  • Excellent separation efficiency even with short sieving times
  • CET Technology for controlled amplitude up to 2,2 mm even with high loads (up to 25 kg)
  • Up to 14 fractions in one sieving operation
  • Free digital adjustment of all process parameters (time, amplitude or sieve acceleration, interval)
  • Memory for up to 9 SOPs
  • Mobile operation panel for comfortable handling
The AS 450 control operates with a three-dimensional sieving motion, which ensures optimum use of the open sieve area and lets the sample move equally over the whole sieving surface.
Bestellinformation: Sieves and all other accessories need to be ordered separately.