Rotating tube divider, PT 200
Lieferant: Retsch GmbH
40.412.0004
RETS40.412.0004EA 6080 EUR
RETS40.412.0004
Rotating tube divider, PT 200
Probenteiler
The PT 200 is a rotating tube divider suitable for representative sampling and exact dividing of larger quantiies of bulk powdered and granular materials.
  • Capable of batch or continuous operation
  • Capable of up to 3 samples of a bulk material - depending on choice of optional bottom cone
  • Adjustable dividing ratios for the three sample collector configurations
  • Quick release sample outlets and easy fastening of bottom cone
Dividing process according to DIN 51701