Debubbler series, Systec®
Lieferant: IDEX RHEODYNE
9000-1549
RHEO9000-1549EA 0 EUR
RHEO9000-1549
Debubbler series, Systec®
Entgaser
Dissolved gases and bubbles in system liquids cause dispense volume anomalies in many instruments, negatively affecting both dispense precision and analytical accuracy. Now you have a choice of components for actively removing bubbles with or without also removing dissolved system gases.
Active debubbler: Removes bubbles in fluid stream before or after the pump.
Degasser/debubbler: Combines vacuum degassing with active bubble removal.
Transfer-line degasser: Removes dissolved gasses during fluid transfer.
* Internal volume in transfer line and in bubble trap.