Wasser ACS, Reagenzqualität ASTM Type I, ASTM Type II Packaged in plastic containers

Lieferant: Ricca Chemical

Synonyme: Wasserstoffoxid, Hydrogenoxid

9150-25
RICC9150-25EA 93.7 EUR
RICC9150-25
Wasser ACS, Reagenzqualität ASTM Type I, ASTM Type II Packaged in plastic containers
Wasser

This product is manufactured by a continuous process that has been validated to produce high purity Water meeting, as a minimum, the specifications below at the time of packaging. This water also meets ASTM Type III and Type IV specifications.

Formel: H₂O
Molecular Weight: 18.02 g/mol
Siedepunkt: 100
Schmelzpunkt: 0
Dichte: 1 g/cm³ (4 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00011332
CAS-Nummer: 7732-18-5
EINECS: 231-791-2
Merck Index: 13,10098

Bestellen

Spezifikation Testergebnisse

Appearance Colorless liquid
Chloride (Cl) max 1 ppb
Conductivity at 25°C (Method: SQCP031) max 0.056 µS/cm
Heavy Metals (as Pb) max 0.01 ppm
Nitrate (NO₃) max 0.4 ppm
Organic Carbon (TOC) max 50 ppb
Oxidizable Substances (Permanganate Retention) To Pass Test
Phosphate (PO₄) max 1.0 ppm
Silicate (as SiO₂) max 3 ppb
Sodium (Na) max 1 ppb
Sulfate (SO₄) max 1.0 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR