Juice Bottle, Clear Glass, TO 43 Round
Lieferant: RIXIUS
1-9900-0250-43
RIXI1-9900-0250-43EA 1770 EUR
RIXI1-9900-0250-43
Juice Bottle, Clear Glass, TO 43 Round
Flaschen Glasflaschen
Juice bottle made of clear glass twist-off thread TO 43 mm.
  • Hydrolytic Class III
  • 250 ml volume capacity
  • Tasteless and odorless
  • Not suitable for carbonation
  • Closures: Twist-off closure
Suitable for hot filling up to max. 95 °C (taking into account the maximum temperature shock of max. 42 °C)
Use: perfect for bottling milk or juices.