Lid for Cans, PP
Lieferant: RIXIUS
8-4100-65
RIXI8-4100-65EA 22.4 EUR
RIXI8-4100-65
Lid for Cans, PP
Verschlüsse Safety Caps und Safety Waste Caps
Screw lids made of white coloured plastic PP.
  • Opening thread 65 mm
  • Without foam insert
  • Complete emptying possible
Suitable for cylindrical wide-neck containers.